Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MCC950 sodium salt 98%

Supplier: AMBEED, INC
Warning

A110681-10MG A110681-1MG A110681-5MG
77233-528EA 28 USD InStock
77233-528 77233-530 77233-532
MCC950 sodium salt 98%
MCC950 sodium salt
Precaución: for Research Use Only


Formula: C₂₀H₂₃N₂NaO₅S
MW: 426.5 g/mol
MDL Number: MFCD30478884
CAS Number: 256373-96-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty