Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butylresorcinol 98%

Supplier: AMBEED, INC
Warning

A110872-10G A110872-10MG A110872-1G A110872-1MG A110872-250MG A110872-25G A110872-50MG A110872-5G
77245-680EA 250 USD InStock
77245-680 77245-682 77245-684 77245-686 77245-688 77245-690 77245-692 77245-694
4-Butylresorcinol 98%
4-Butylresorcinol
Precaución: for Research Use Only


Formula: C₁₀H₁₄O₂
MDL Number: MFCD01684800
CAS Number: 18979-61-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty