Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,7-Dibromo-6-methyl-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-indazole 97%

Supplier: AMBEED, INC
Warning

A110937-100G A110937-10G A110937-1G A110937-25G A110937-5G
77233-550EA 272 USD InStock
77233-550 77233-552 77233-554 77233-556 77233-558
5,7-Dibromo-6-methyl-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-indazole 97%
5,7-Dibromo-6-methyl-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-indazole
Precaución: for Research Use Only


Formula: C₁₄H₂₀Br₂N₂OSi
MDL Number: MFCD30342443
CAS Number: 1973503-49-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty