Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-(trifluoromethyl)phenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A112072-10G A112072-1G A112072-25G A112072-5G
77257-702EA 23 USD InStock
77257-702 77257-704 77257-706 77257-708
2-Fluoro-3-(trifluoromethyl)phenylboronic acid 97%
2-Fluoro-3-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₅BF₄O₂
MDL Number: MFCD07784336
CAS Number: 157834-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty