Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-3-methoxybenzoicacid 98%

Supplier: AMBEED, INC
Warning

A112659-10G A112659-1G A112659-25G A112659-5G
77284-252EA 58 USD InStock
77284-252 77284-254 77284-256 77284-258
4-Benzyloxy-3-methoxybenzoicacid 98%
4-Benzyloxy-3-methoxybenzoicacid
Precaución: for Research Use Only


Formula: C₁₅H₁₄O₄
MW: 258.2693 g/mol
MDL Number: MFCD00183281
CAS Number: 1486-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty