Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

EPZ015866 (GSK591) ≥98%

Supplier: AMBEED, INC
Warning

A112992-10MG A112992-1MG A112992-25MG A112992-5MG
77233-684EA 33 USD InStock
77233-684 77233-686 77233-688 77233-690
EPZ015866 (GSK591) ≥98%
EPZ015866 (GSK591)
Precaución: for Research Use Only


Formula: C₂₂H₂₈N₄O₂
MDL Number: MFCD30187524
CAS Number: 1616391-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty