Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Fluoro-3'-methylacetophenone 95%

Supplier: AMBEED, INC
Warning

A113129-100G A113129-10G A113129-1G A113129-25G A113129-5G
77271-014EA 366.01 USD InStock
77271-014 77271-016 77271-018 77271-020 77271-022
4'-Fluoro-3'-methylacetophenone 95%
4'-Fluoro-3'-methylacetophenone
Precaución: for Research Use Only


Formula: C₉H₉FO
MW: 152.17
MDL Number: MFCD04972879
CAS Number: 369-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty