Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Benzyloxyaniline 98%

Supplier: AMBEED, INC
Warning

A113442-100G A113442-10G A113442-1G A113442-25G A113442-5G
77271-038EA 47 USD InStock
77271-038 77271-040 77271-042 77271-044 77271-046
3-Benzyloxyaniline 98%
3-Benzyloxyaniline
Precaución: for Research Use Only


Formula: C₁₃H₁₃NO
MDL Number: MFCD00007784
CAS Number: 1484-26-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty