Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-chloronicotinate 98%

Supplier: AMBEED, INC
Warning

A113753-100G A113753-10G A113753-500G A113753-5G
77233-774EA 42 USD InStock
77233-774 77233-776 77233-778 77233-780
Ethyl-2-chloronicotinate 98%
Ethyl-2-chloronicotinate
Precaución: For research use only.


Formula: C₈H₈ClNO₂
MW: 185.61
MDL Number: MFCD00674129
CAS Number: 1452-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty