Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-1-fluorobenzene 95%

Supplier: AMBEED, INC
Warning

A114167-100G A114167-25G
77284-342EA 64 USD InStock
77284-342 77284-344 77284-346 77284-348
2,4-Dibromo-1-fluorobenzene 95%
2,4-Dibromo-1-fluorobenzene
Precaución: for Research Use Only


Formula: C₆H₃Br₂F
MW: 253.9 g/mol
MDL Number: MFCD00000283
CAS Number: 1435-53-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty