Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

KRIBB11 98%

Supplier: AMBEED, INC
Warning

A114945-10MG A114945-1MG A114945-25MG A114945-5MG A114945-100MG A114945-250MG A114945-50MG
77257-860EA 42 USD InStock
77257-860 77257-862 77257-864 77257-866 77747-842 77747-843 77747-844
KRIBB11 98%
KRIBB11
Precaución: for Research Use Only


Formula: C₁₃H₁₂N₆O₂
MDL Number: MFCD30182306
CAS Number: 342639-96-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty