Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-methylbenzo[d]thiazol-2-amine 97%

Supplier: AMBEED, INC
Warning

A115928-1G A115928-250MG A115928-5G
77245-912EA 88 USD InStock
77245-912 77245-914 77245-916
5-Bromo-4-methylbenzo[d]thiazol-2-amine 97%
5-Bromo-4-methylbenzo[d]thiazol-2-amine


Formula: C₈H₇BrN₂S
MDL Number: MFCD23705662
CAS Number: 103873-80-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty