Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-2-Phenylcyclopropylamine hydrochloride 97%

Supplier: AMBEED, INC
Danger

A116070-100MG A116070-10MG A116070-250MG A116070-50MG A116070-5G A116070-1G A116070-25G
77245-926EA 11 USD InStock
77245-926 77245-928 77245-930 77245-932 77566-098 77745-857 77745-858
trans-2-Phenylcyclopropylamine hydrochloride 97%
trans-2-Phenylcyclopropylamine hydrochloride
Precaución: For research use only.


Formula: C₉H₁₂ClN
MDL Number: MFCD00063602
CAS Number: 1986-47-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty