Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indol-3-ylhydrosulfide 98%

Supplier: AMBEED, INC
Warning

A116814-100MG A116814-1G A116814-250MG
77271-252EA 40 USD InStock
77271-252 77271-254 77271-256
1H-Indol-3-ylhydrosulfide 98%
1H-Indol-3-ylhydrosulfide
Precaución: for Research Use Only


Formula: C₈H₇NS
MW: 149.22 g/mol
MDL Number: MFCD00152110
CAS Number: 480-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty