Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 2-(4-nitrobenzyl)malonate 96%

Supplier: AMBEED, INC
Warning

A116883-100MG A116883-1G A116883-250MG
77257-964EA 13 USD InStock
77257-964 77257-966 77257-968
Diethyl 2-(4-nitrobenzyl)malonate 96%
Diethyl 2-(4-nitrobenzyl)malonate
Precaución: for Research Use Only


Formula: C₁₄H₁₇NO₆
MDL Number: MFCD00024804
CAS Number: 7598-70-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty