Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CP-724714 98+%

Supplier: AMBEED, INC
Warning

A116887-10MG A116887-1MG A116887-5MG
77233-940EA 52 USD InStock
77233-940 77233-942 77233-944
CP-724714 98+%
CP-724714
Precaución: for Research Use Only


Formula: C₂₇H₂₇N₅O₃
MDL Number: MFCD11983048
CAS Number: 383432-38-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty