Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-4-isoxazolecarbaldehyde 98%

Supplier: AMBEED, INC
Warning

A117227-1G A117227-25G A117227-5G
77284-614EA 10 USD InStock
77284-614 77284-616 77284-618
3,5-Dimethyl-4-isoxazolecarbaldehyde 98%
3,5-Dimethyl-4-isoxazolecarbaldehyde
Precaución: for Research Use Only


Formula: C6H7NO2
MW: 125.13 g/mol
MDL Number: MFCD02681977
CAS Number: 54593-26-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty