Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrimidin-2-ylmethanamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A117604-10G A117604-1G A117604-25G A117604-5G A117604-100G
77257-990EA 12 USD InStock
77257-990 77257-992 77257-994 77257-996 77745-870
Pyrimidin-2-ylmethanamine hydrochloride 98%
Pyrimidin-2-ylmethanamine hydrochloride
Precaución: for Research Use Only


Formula: C5H8ClN3
MW: 145.59 g/mol
MDL Number: MFCD08062208
CAS Number: 372118-67-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty