Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis[4-(3-aminophenoxy)phenyl]sulfone 98%

Supplier: AMBEED, INC
Warning

A118040-100G A118040-10G A118040-25G A118040-5G
77284-646EA 43 USD InStock
77284-646 77284-648 77284-650 77284-652
Bis[4-(3-aminophenoxy)phenyl]sulfone 98%
Bis[4-(3-aminophenoxy)phenyl]sulfone
Precaución: for Research Use Only


Formula: C₂₄H₂₀N₂O₄S
MDL Number: MFCD00054738
CAS Number: 30203-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty