Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4'-Biphthalic anhydride 98%

Supplier: AMBEED, INC
Warning

A118115-100G A118115-1G A118115-25G A118115-500G A118115-5G
77245-994EA 132 USD InStock
77245-994 77245-996 77245-998 77246-000 77246-002
3,4'-Biphthalic anhydride 98%
3,4'-Biphthalic anhydride
Precaución: for Research Use Only


Formula: C₁₆H₆O₆
MDL Number: MFCD09030654
CAS Number: 36978-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty