Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-fluoro-2-iodobenzene 98%

Supplier: AMBEED, INC
Warning

A118900-100G A118900-10G A118900-1G A118900-25G
77284-704EA 46 USD InStock
77284-704 77284-706 77284-708 77284-710
4-Bromo-1-fluoro-2-iodobenzene 98%
4-Bromo-1-fluoro-2-iodobenzene
Precaución: for Research Use Only


Formula: C₆H₃BrFI
MDL Number: MFCD07368751
CAS Number: 116272-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty