Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PPM1D phosphatase inhibitor ≥98%

Supplier: AMBEED, INC
Warning

A119848-10MG A119848-50MG A119848-5MG
77234-044EA 60 USD InStock
77234-044 77234-046 77234-048 77234-050
PPM1D phosphatase inhibitor ≥98%
PPM1D phosphatase inhibitor
Precaución: for Research Use Only


Formula: C₁₅H₁₂OS₂
MDL Number: MFCD00121546
CAS Number: 176957-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty