Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-m-anisaldehyde 98%

Supplier: AMBEED, INC
Warning

A119953-100G A119953-10G A119953-25G A119953-500G A119953-1000G
77258-068EA 12 USD InStock
77258-068 77258-070 77258-072 77258-074 77747-874
2-Hydroxy-m-anisaldehyde 98%
2-Hydroxy-m-anisaldehyde
Precaución: for Research Use Only


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 265
MDL Number: MFCD00003322
CAS Number: 148-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty