Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Liproxstatin-1 98%

Supplier: AMBEED, INC
Warning

A120662-100MG A120662-10MG A120662-1MG A120662-2MG A120662-50MG A120662-5MG A120662-250MG
77246-092EA 448 USD InStock
77246-092 77246-094 77246-096 77246-098 77246-100 77246-102 77745-888
Liproxstatin-1 98%
Liproxstatin-1
Precaución: for Research Use Only


Formula: C₁₉H₂₁ClN₄
MDL Number: MFCD14948691
CAS Number: 950455-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty