Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)-2-pyrazolin-5-one 95%

Supplier: AMBEED, INC
Warning

A120887-10G A120887-1G A120887-25G A120887-5G
77271-490EA 85 USD InStock
77271-490 77271-492 77271-494 77271-496
3-(Trifluoromethyl)-2-pyrazolin-5-one 95%
3-(Trifluoromethyl)-2-pyrazolin-5-one
Precaución: for Research Use Only


Formula: C₄H₃F₃N₂O
MDL Number: MFCD00052748
CAS Number: 401-73-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty