Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Leucine 97%

Supplier: AMBEED, INC
Warning

A121112-100G A121112-25G
77271-502EA 85 USD InStock
77271-502 77271-504
DL-Leucine 97%
DL-Leucine
Precaución: for Research Use Only


Formula: C₆H₁₃NO₂
MDL Number: MFCD00063087
CAS Number: 328-39-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty