Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1-fluoro-3-nitrobenzene 98%

Supplier: AMBEED, INC
Warning

A121310-10G A121310-1G A121310-25G A121310-5G
77284-822EA 70 USD InStock
77284-822 77284-824 77284-826 77284-828
2-Chloro-1-fluoro-3-nitrobenzene 98%
2-Chloro-1-fluoro-3-nitrobenzene
Precaución: for Research Use Only


Formula: C₆H₃ClFNO₂
MDL Number: MFCD04112549
CAS Number: 21397-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty