Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 12-bromododecanoate 97%

Supplier: AMBEED, INC
Warning

A121528-100MG A121528-1G A121528-250MG A121528-25G A121528-5G
77234-132EA 7 USD InStock
77234-132 77234-134 77234-136 77234-138 77234-140
Ethyl 12-bromododecanoate 97%
Ethyl 12-bromododecanoate
Precaución: for Research Use Only


Formula: C₁₄H₂₇BrO₂
MDL Number: MFCD30470568
CAS Number: 72338-48-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty