Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diphenylpyridine 98%

Supplier: AMBEED, INC
Warning

A121676-100MG A121676-250MG A121676-1G
77246-134EA 36 USD InStock
77246-134 77246-136 77745-894
2,4-Diphenylpyridine 98%
2,4-Diphenylpyridine
Precaución: for Research Use Only


Formula: C₁₇H₁₃N
MDL Number: MFCD00972966
CAS Number: 26274-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty