Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxybenzhydrazide 95%

Supplier: AMBEED, INC
Warning

A122191-100G A122191-1G A122191-250MG A122191-25G A122191-5G
77246-148EA 347 USD InStock
77246-148 77246-150 77246-152 77246-154 77246-156
2-Ethoxybenzhydrazide 95%
2-Ethoxybenzhydrazide
Precaución: for Research Use Only


Formula: C₉H₁₂N₂O₂
MDL Number: MFCD00017050
CAS Number: 21018-13-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty