Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methylthio)pyrimidin-4-amine 98%

Supplier: AMBEED, INC
Warning

A122372-100MG A122372-1G A122372-250MG
77246-162EA 15 USD InStock
77246-162 77246-164 77246-166
2-(Methylthio)pyrimidin-4-amine 98%
2-(Methylthio)pyrimidin-4-amine
Precaución: for Research Use Only


Formula: C₅H₇N₃S
MW: 141.1942 g/mol
MDL Number: MFCD02091098
CAS Number: 2183-66-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty