Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thenoic acid 98%

Supplier: AMBEED, INC
Warning

A122589-100G A122589-10G A122589-1G A122589-25G A122589-500G A122589-5G
77284-890EA 70 USD InStock
77284-890 77284-892 77284-894 77284-896 77284-898 77284-900
3-Thenoic acid 98%
3-Thenoic acid
Precaución: for Research Use Only


Formula: C₅H₄O₂S
MW: 128.15 g/mol
Melting Pt: 137
MDL Number: MFCD00005467
CAS Number: 88-13-1
Merck Index: 12,09415
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty