Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-2-methoxypyridine 98%

Supplier: AMBEED, INC
Warning

A122621-10G A122621-1G A122621-250MG A122621-25G A122621-5G
77258-134EA 48 USD InStock
77258-134 77258-136 77258-138 77258-140 77258-142
5-Iodo-2-methoxypyridine 98%
5-Iodo-2-methoxypyridine
Precaución: for Research Use Only


Formula: C₆H₆INO
MDL Number: MFCD07781180
CAS Number: 13472-61-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty