Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-2-methylpyrimidine 98%

Supplier: AMBEED, INC
Warning

A122636-1G A122636-250MG A122636-25G
77284-910EA 14 USD InStock
77284-910 77284-912 77284-914
4-Hydroxy-2-methylpyrimidine 98%
4-Hydroxy-2-methylpyrimidine
Precaución: for Research Use Only


Formula: C₅H₆N₂O
MDL Number: MFCD00234103
CAS Number: 19875-04-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty