Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-6-propoxypyridazine 97%

Supplier: AMBEED, INC
Warning

A123517-1G A123517-25G
77271-658EA 40.17 USD InStock
77271-658 77271-660 77271-662
3-Chloro-6-propoxypyridazine 97%
3-Chloro-6-propoxypyridazine
Precaución: for Research Use Only


Formula: C₇H₉ClN₂O
MDL Number: MFCD08460980
CAS Number: 5788-60-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty