Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

S 3304 97%

Supplier: AMBEED, INC
Warning

A123766-10MG A123766-1MG A123766-25MG A123766-50MG A123766-5MG
77258-158EA 55 USD InStock
77258-158 77258-160 77258-162 77258-164 77258-166
S 3304 97%
S 3304
Precaución: for Research Use Only


Formula: C₂₄H₂₀N₂O₄S₂
MDL Number: MFCD22200275
CAS Number: 203640-27-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty