Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Kojic acid dipalmitate 97%

Supplier: AMBEED, INC
Warning

A123792-10G A123792-25G
77271-686EA 11 USD InStock
77271-686 77271-688
Kojic acid dipalmitate 97%
Kojic acid dipalmitate
Precaución: for Research Use Only


Formula: C₃₈H₆₆O₆
MDL Number: MFCD03840587
CAS Number: 79725-98-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty