Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Nitrophenyl)glyoxylic acid 95%

Supplier: AMBEED, INC
Warning

A125734-10G A125734-1G A125734-250MG
77285-106EA 523 USD InStock
77285-106 77285-108 77285-110
(4-Nitrophenyl)glyoxylic acid 95%
(4-Nitrophenyl)glyoxylic acid
Precaución: for Research Use Only


Formula: C₈H₅NO₅
MDL Number: MFCD00051744
CAS Number: 14922-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty