Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-ethoxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A125906-10G A125906-5G
77271-784EA 50 USD InStock
77271-784 77271-786
3-Bromo-4-ethoxybenzaldehyde 97%
3-Bromo-4-ethoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₉BrO₂
MDL Number: MFCD00672941
CAS Number: 108373-05-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty