Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium pamidronate 98% (contains H₂O)

Supplier: AMBEED, INC
Warning

A126151-10G A126151-1G A126151-250MG A126151-25G A126151-5G
77271-788EA 704.01 USD InStock
77271-788 77271-790 77271-792 77271-794 77271-796
Disodium pamidronate 98% (contains H₂O)
Disodium pamidronate
Precaución: for Research Use Only


Formula: C₃H₉NO₇P₂Na₂
MW: 279.03 g/mol
MDL Number: MFCD11045314
CAS Number: 57248-88-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty