Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyclopropyl-1H-pyrazole-4-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A126172-250MG A126172-100MG
77234-318EA 30 USD InStock
77234-318 77745-916
1-Cyclopropyl-1H-pyrazole-4-carbaldehyde 97%
1-Cyclopropyl-1H-pyrazole-4-carbaldehyde
Precaución: For research use only.


Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD19687735
CAS Number: 1082066-00-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty