Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-4-tert-butylphenol 95%

Supplier: AMBEED, INC
Warning

A126383-1G A126383-250MG A126383-5G
77246-346EA 6 USD InStock
77246-346 77246-348 77246-350
2,6-Dibromo-4-tert-butylphenol 95%
2,6-Dibromo-4-tert-butylphenol
Precaución: For research use only.


Formula: C₁₀H₁₂Br₂O
MDL Number: MFCD00152389
CAS Number: 98-22-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty