Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Morpholin-4-ylmethyl)benzonitrile 97%

Supplier: AMBEED, INC
Danger

A126658-1G A126658-250MG A126658-5G
77285-138EA 34 USD InStock
77285-138 77285-140 77285-142
4-(Morpholin-4-ylmethyl)benzonitrile 97%
4-(Morpholin-4-ylmethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₁₂H₁₄N₂O
MW: 202.2500 g/mol
MDL Number: MFCD00454271
CAS Number: 37812-51-4
UN: 2923
ADR: 8,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty