Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cinnamamide 97% (Predominantly trans)

Supplier: AMBEED, INC
Warning

A126745-100G A126745-10G A126745-500G A126745-25G
77271-818EA 32 USD InStock
77271-818 77271-820 77271-822 77298-148
Cinnamamide 97% (Predominantly trans)
Cinnamamide
Precaución: for Research Use Only


Formula: C₉H₉NO
MDL Number: MFCD00008033
CAS Number: 621-79-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty