Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Difluoro-5-iodobenzene 98%

Supplier: AMBEED, INC
Warning

A127175-10G A127175-1G A127175-25G A127175-5G
77285-148EA 15 USD InStock
77285-148 77285-150 77285-152 77285-154
1,3-Difluoro-5-iodobenzene 98%
1,3-Difluoro-5-iodobenzene
Precaución: for Research Use Only


Formula: C₆H₃F₂I
MW: 239.99 g/mol
MDL Number: MFCD03094155
CAS Number: 2265-91-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty