Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Bromo-3-chloropropiophenone 98%

Supplier: AMBEED, INC
Warning

A127908-100G A127908-10G A127908-25G A127908-5G
77258-330EA 148 USD InStock
77258-330 77258-332 77258-334 77747-913
4'-Bromo-3-chloropropiophenone 98%
4'-Bromo-3-chloropropiophenone
Precaución: for Research Use Only


Formula: C₉H₈BrClO
MW: 247.52 g/mol
Boiling Pt: 301
MDL Number: MFCD00018979
CAS Number: 31736-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty