Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,6-xylenol 95%

Supplier: AMBEED, INC
Warning

A128054-100G A128054-10G A128054-25G A128054-5G
77285-170EA 84 USD InStock
77285-170 77285-172 77285-174 77285-176
4-Bromo-2,6-xylenol 95%
4-Bromo-2,6-xylenol
Precaución: for Research Use Only


Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 259
Melting Pt: 79
MDL Number: MFCD00002314
CAS Number: 2374-05-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty