Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GlyH-101 98+%

Supplier: AMBEED, INC
Warning

A128948-10MG A128948-1MG A128948-2MG A128948-50MG A128948-5MG
77234-384EA 32 USD InStock
77234-384 77234-386 77234-388 77234-390 77234-392
GlyH-101 98+%
GlyH-101
Precaución: for Research Use Only


Formula: C₁₉H₁₅Br₂N₃O₃
MW: 493.15 g/mol
MDL Number: MFCD01532954
CAS Number: 328541-79-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty