Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4-Trihydroxybenzophenone 98%

Supplier: AMBEED, INC
Warning

A129271-1000G A129271-100G A129271-25G A129271-500G A129271-5G
77246-464EA 305.01 USD InStock
77246-464 77246-466 77246-468 77246-470 77246-472 77246-474
2,3,4-Trihydroxybenzophenone 98%
2,3,4-Trihydroxybenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₁₀O₄
MDL Number: MFCD00009996
CAS Number: 1143-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty