Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Metacetamol 98%

Supplier: AMBEED, INC
Warning

A129748-100G A129748-10G A129748-25G A129748-5G A129748-500G
77271-948EA 51 USD InStock
77271-948 77271-950 77271-952 77271-954 77747-926
Metacetamol 98%
Metacetamol
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MDL Number: MFCD00002263
CAS Number: 621-42-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty